Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA018419

Sigma-Aldrich

Anti-POLDIP3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-46 kDa DNA polymerase delta interaction protein, Anti-Polymerase delta-interacting protein 3, Anti-p46

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human, mouse

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

SLDELIRKRGAAAKGRLNARPGVGGVRSRVGIQQGLLSQSTRTATFQQRFDARQKIGLSDARLKLGVKDAREKLLQKDARFRIKGKVQDAR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... POLDIP3(84271)

Related Categories

General description

The gene POLDIP3 (polymerase δ-interacting protein 3) is mapped to human chromosome 22q13.2. It is also popularly referred as SKAR. POLDIP3 is expressed ubiquitously, with highest levels in brain, heart, skeletal muscle, and kidney. The protein is mainly localized in the nucleus and exhibit a punctuate pattern. However, POLDIP3 can shuttle between the nucleus and the cytoplasm.

Immunogen

Polymerase delta-interacting protein 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Polymerase δ-interacting protein 3 (POLDIP3) is deposited at the exon-junction complex and recruits S6K1 (ribosomal protein S6 kinase β-1) to the mRNA. POLDIP3 and S6K1 together are important to increase the translation efficiency of spliced mRNA. S6K1 lies downstream of mTOR (mammalian target of rapamycin) and PI3K (phosphoinositides 3-kinase), which are involved in cell growth promotion and cell cycle progression. Absence of S6K1 or S6K1 interacting partner POLDIP3 decreases cell size. IFN-α (interferon-α) induces phosphorylation of POLDIP3, thereby enhancing latter interaction with the eukaryotic initiation factor 4G. This pathway is important for regulation of mRNA translation of IFN-sensitive genes and induction of IFN-α biological response. POLDIP3 also interacts with the p50 subunit of DNA polymerase δ and enhancer of rudimentary protein.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74560

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julio Avila et al.
Journal of autoimmune diseases, 5, 3-3 (2008-07-16)
Systemic vasculitides constitute a heterogeneous group of diseases of autoimmunological origin characterized by inflammation of blood vessels and antibodies that react against autoantigens in a process that ultimately affects blood vessel walls. An important number of these patients present kidney
Barbara Kroczynska et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(31), 11377-11382 (2014-07-23)
We provide evidence that S6 kinase 1 (S6K1) Aly/REF-like target (SKAR) is engaged in IFN-α signaling and plays a key role in the generation of IFN responses. Our data demonstrate that IFN-α induces phosphorylation of SKAR, which is mediated by
Celeste J Richardson et al.
Current biology : CB, 14(17), 1540-1549 (2004-09-03)
The mammalian target of rapamycin (mTOR) and phosphatidylinositol 3-kinase (PI3K) signaling pathways promote cell growth and cell cycle progression in response to nutritional, energy, and mitogenic cues. In mammalian cells, the ribosomal protein S6 kinases, S6K1 and S6K2, lie downstream
Amelia Smyk et al.
The FEBS journal, 273(20), 4728-4741 (2006-09-21)
Enhancer of rudimentary (ER) is a small protein that has a unique amino acid sequence and structure. Its highly conserved gene has been found in all eukaryotic kingdoms with the exception of fungi. ER was proposed to be involved in
Xiaoju Max Ma et al.
Cell, 133(2), 303-313 (2008-04-22)
Different protein complexes form on newly spliced mRNA to ensure the accuracy and efficiency of eukaryotic gene expression. For example, the exon junction complex (EJC) plays an important role in mRNA surveillance. The EJC also influences the first, or pioneer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service