Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA007832

Sigma-Aldrich

Anti-PRMT3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3, Anti-Protein arginine N-methyltransferase 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

TFSHCKSEHQFNIDSMVHKHGLEFYGYIKLINFIRLKNPTVEYMNSIYNPVPWEKEEYLKPVLEDDLLLQFDVEDLYEPVSVPFSYPNGLSENTSVVEKLKHMEARALSAEAALARAREDLQKMKQFAQDFVMHTDVRTCSSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRMT3(10196)

General description

PRMT3 (protein arginine methyltransferase 3) gene encodes a member of the protein arginine methyltransferase (PRMT) family. It contains an N-terminal acidic amino acid-rich (NAR) domain that comprises of two functional motifs, a putative C2H2 zinc finger motif and a tyrosine phosphorylation consensus sequence. The C-terminus contains a putative methyltransferase domain. It is a monomeric protein mostly localized to the cytoplasm.

Immunogen

Protein arginine N-methyltransferase 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PRMT3 (protein arginine methyltransferase 3) gene encodes a methyltransferase that catalyzes the methylation of guanidino nitrogens of arginyl residues of proteins to form asymmetric dimethylarginine. The small 40S ribosomal protein S2 (rpS2) forms a zinc-finger-dependent substrate of mammalian PRMT3. It is important in 80S ribosome maturation and in protein synthesis. It balances the cellular levels of ribosomal subunits.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70288

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

François Bachand et al.
The EMBO journal, 23(13), 2641-2650 (2004-06-04)
The mammalian protein arginine methyltransferase 3 (PRMT3) catalyzes the formation of asymmetric (type I) dimethylarginine in vitro. As yet, natural substrates and cellular pathways modulated by PRMT3 remain unknown. Here, we have identified an ortholog of PRMT3 in fission yeast.
Shingo Miyata et al.
Brain research, 1352, 11-20 (2010-07-22)
Protein arginine N-methyltransferase 3 (PRMT3) is a cytoplasmic enzyme that utilizes S-adenosyl-L-methionine (AdoMet) to methylate specific proteins, most of which contain GAR (glycine-arginine rich) motifs. PRMT3 has been shown to play a role in the proper maturation of the 80S
X Zhang et al.
The EMBO journal, 19(14), 3509-3519 (2000-07-19)
Protein arginine methylation has been implicated in signal transduction, nuclear transport and transcription regulation. Protein arginine methyltransferases (PRMTs) mediate the AdoMet-dependent methylation of many proteins, including many RNA binding proteins involved in various aspects of RNA processing and/or transport. Here
J Tang et al.
The Journal of biological chemistry, 273(27), 16935-16945 (1998-06-27)
Methylation is one of the many post-translational modifications that modulate protein function. Although asymmetric NG,NG-dimethylation of arginine residues in glycine-arginine-rich domains of eucaryotic proteins, catalyzed by type I protein arginine N-methyltransferases (PRMT), has been known for some time, members of
Rafal Swiercz et al.
The Biochemical journal, 386(Pt 1), 85-91 (2004-10-12)
PRMT3 (protein arginine methyltransferase 3) is one of four type I arginine methyltransferases that catalyse the formation of asymmetric dimethylarginine. PRMT3 is unique in that its N-terminus harbours a C2H2 zinc-finger domain that is proposed to confer substrate specificity. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service