Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA004198

Sigma-Aldrich

Anti-NCOA6 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ASC-2 antibody produced in rabbit, Anti-Activating signal cointegrator 2 antibody produced in rabbit, Anti-Amplified in breast cancer protein 3 antibody produced in rabbit, Anti-Cancer-amplified transcriptional coactivator ASC-2 antibody produced in rabbit, Anti-Nuclear receptor coactivator 6 antibody produced in rabbit, Anti-Peroxisome proliferator-activated receptor-interacting protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
clone:
polyclonal
application:
IF
IHC
species reactivity:
human
technique(s):
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200
citations:
2

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

NQNVQHAGGQGAGPPQNQMQVSHGPPNMMQPSLMGIHGNMNNQQAGTSGVPQVNLSNMQGQPQQGPPSQLMGMHQQIVPSQGQMVQQQGTLNPQNPMILSRAQLMPQGQMMVNPPSQNLGPSPQRMTPPKQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NCOA6(23054)

General description

NCOA6 (Nuclear receptor coactivator 6) encodes a novel human nuclear receptor coactivator protein. Its modular structure is composed of a large intrinsic glutamine-rich activation domain and a single functional LXXLL motif. LXXLL motif helps to associate with the members of the steroid hormone and thyroid hormone/retinoid receptor subfamilies. In humans, it consists of a potent N-terminal activation domain (AD1) and a second activation domain (AD2). It is copiously expressed in several reproductive organs such as testis, prostate, ovary and also in brain.

Immunogen

Nuclear receptor coactivator 6 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NCOA6 (Nuclear receptor coactivator 6) interacts with nuclear receptors in ligand-dependent or ligand-enhanced dependent manner to regulate nuclear receptor mediated mammalian gene expression. The activation domain helps to increase the transcriptional activity of several nuclear receptors. It forms dimer by binding to the liganded receptors which leads to a structural change in NRC activation. It has also been reported that NCOA6 highly influences the binding of CREB-binding protein (CBP) during transcriptional activity of nuclear hormone receptors.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86732

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Mahajan et al.
Molecular and cellular biology, 20(14), 5048-5063 (2000-06-24)
We describe the cloning and characterization of a new family of nuclear receptor coregulators (NRCs) which modulate the function of nuclear hormone receptors in a ligand-dependent manner. NRCs are expressed as alternatively spliced isoforms which may exhibit different intrinsic activities
F Caira et al.
The Journal of biological chemistry, 275(8), 5308-5317 (2000-02-22)
Ligand-induced transcriptional activation of gene expression by nuclear receptors is dependent on recruitment of coactivators as intermediary factors. The present work describes the cloning and characterization of RAP250, a novel human nuclear receptor coactivator. The results of in vitro and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service