Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-5482-11

HiTrap® LambdaFabSelect

pack of 5 × 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

packaging

pack of 5 × 1 mL

manufacturer/tradename

Cytiva

column I.D.

7 mm

matrix

highly cross-linked agarose

particle size

75 μm

cleaning

2-12

working range

3-10

storage temp.

2-8°C

General description

LambdaFabSelect is an affinity chromatography media, designed for the purification of lambda Fab fragments, with high binding capacity, purity, and yield.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

HiTrap is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service