Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G21110-HH

Sigma-Aldrich

DMT-2′O-Methyl-rG(ib) Phosphoramidite

Synonym(s):

DMT-2′-O-Me-rG(ib) amidite, N-(2-methyl-1-oxopropyl)-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-methyl-guanosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H56N7O9P
CAS Number:
Molecular Weight:
869.94
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

non-animal source (no BSE/TSE risk)

Quality Level

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

packaging

pkg of 1 × 100 g (G211111-01)
pkg of 1 × 500 g G211151-01

technique(s)

oligo synthesis: suitable

impurities

≤0.1% single unspecified Impurity (reversed phase HPLC)
≤0.3% mG2 (reversed phase HPLC, Hydrolysate)
≤0.3% mG3 (reversed phase HPLC, DMT-rG(ib)me)
≤0.3 wt. % water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤1.0% mG1 (reversed phase HPLC, DMT-rG(ib)me-DMT)
≤3 wt. % residual Solvent content

color

white to off-white

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

nucleoside profile

base: guanosine
base protecting group: isobutyryl
2' protecting group: methyl
5' protecting group: DMT
deprotection: standard

storage temp.

2-8°C

SMILES string

CO[C@@H]1[C@H](OP(OCCC#N)N(C(C)C)C(C)C)[C@@H](COC(c2ccccc2)(c3ccc(OC)cc3)c4ccc(OC)cc4)O[C@H]1n5cnc6C(=O)NC(NC(=O)C(C)C)=Nc56

InChI

1S/C45H56N7O9P/c1-28(2)41(53)49-44-48-40-37(42(54)50-44)47-27-51(40)43-39(57-9)38(61-62(59-25-13-24-46)52(29(3)4)30(5)6)36(60-43)26-58-45(31-14-11-10-12-15-31,32-16-20-34(55-7)21-17-32)33-18-22-35(56-8)23-19-33/h10-12,14-23,27-30,36,38-39,43H,13,25-26H2,1-9H3,(H2,48,49,50,53,54)/t36-,38-,39-,43-,62?/m1/s1

InChI key

IRRDHRZUOZNWDJ-MLLDKZSOSA-N

Looking for similar products? Visit Product Comparison Guide

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service