EP951020648
Eppendorf® twin.tec PCR Plates 96
Optimized rim for barcodes, skirt, well volume 150 μL, blue, pkg of 300 ea
All Photos(1)
About This Item
material
blue
conical bottom
polypropylene wells
sterility
non-sterile
feature
lid: no
skirt
packaging
pkg of 300 ea
well volume
150 μL
suitability
suitable for PCR
General description
Eppendorf® twin.tec PCR Plate 96, skirted, 150 µL, PCR clean, blue, 300 plates
- Optimized rim for barcodes and automated work processes
- Within SBS footprint recommendations (127.76 × 85.47 mm ±0.25 mm)
- Stackable
- Eight holes in the skirt aid plate positioning and removal from the thermal cycler block
- Low profile design enhances efficiency of PCR and enables the highest efficiency for small sample volumes
- 150 µL max. well volume (when used with cap strips)
- Available with barcode (upon request), for skirted and semi-skirted only
Features and Benefits
- Semi-skirted for label and barcode
- Fits most thermal cyclers
- 250 µL max. well volume (when used with cap strips)
- Improved well-to-well tolerance
- Available with barcode (upon request), for skirted and semi-skirted only
Legal Information
Eppendorf is a registered trademark of Eppendorf AG