Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP5490048008

Eppendorf® Fixed-Angle Rotor F-45-22-17

to hold, 22 x 5 mL conical tubes, used with Concentrator Plus

Synonym(s):

Centrifuge rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909

feature

to hold 22 x 5 mL conical tubes

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5490048008

compatibility

used with Concentrator Plus

General description

Rotor F-45-22-17, 22 places for 5 mL conical tubes

Application

CENTRIFUGATION

Features and Benefits

Max. load: 22 x 5,0 mLTube Options: Snap caps and screw capsTube diameter: 17 mmTube height: 59.4 mm - 67.4 mmMax. g-force (rcf): 242 × gMax. rotational speed (rpm): 1,400 rpmMax. load per bore: 10 g (tube and sample)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service