Key Documents
EP5427734007
Eppendorf® Adapter
holds 1 x 7-15 mL, bore size (large), pack of 2 ea
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
holds 1 x 7-15 mL
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5427734007
technique(s)
centrifugation: suitable
bore size
(large)
compatibility
for use with Rotor F-35-6-30 (large rotor bore)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)