Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMU087951

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Frzb

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCGAACGTTGCAAATGTAAGCCTGTCAGAGCTACACAGAAGACCTATTTCCGGAACAATTACAACTATGTCATCCGGGCTAAAGTTAAAGAGGTAAAGATGAAATGTCATGATGTGACCGCCGTTGTGGAAGTGAAGGAAATTCTAAAGGCATCACTGGTAAACATTCCAAGGGACACCGTCAATCTTTATACCACCTCTGGCTGCCTCTGTCCTCCACTTACTGTCAATGAGGAATATGTCATCATGGGCTATGAAGACGAGGAACGTTCCAGGTTACTCTTGGTAGAAGGCTCTATAGCTGAGAAGTGGAAGGATCGGCTTGGTAAGAAAGTCAAGCGCTGGGATATGAAACTCCGACACCTTGGACTGGGTAAAACTGATGCTAGCGATTCCACTCAGAATCAGAAGTCTGGCAGGAACTCTAATCCCCGGCCAGCACGCAGCTAAATCCTGAAATGTAAAAGGCCACACCCACGGACTCCCTTCTAAGACTGGCGCTGCTGGACTAACAAAGGAAAACCGCACAG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julie J G Kephart et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 21(21), 4868-4880 (2015-06-14)
Rhabdomyosarcoma (RMS) is a soft tissue sarcoma associated with the skeletal muscle lineage. Of the two predominant subtypes, known as embryonal (eRMS) and alveolar (aRMS), aRMS has the poorer prognosis, with a five-year survival rate of <50%. The majority of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service