Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMU061601

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Mylk

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACCCATGGCTGATGAAAGACACCAAGAACATGGAGGCCAAGAAGCTGTCCAAGGACCGGATGAAGAAATACATGGCAAGAAGGAAGTGGCAGAAAACGGGCAATGCTGTGAGAGCCATCGGGAGACTGTCCTCTATGGCAATGATCTCAGGGCTCAGTGGCAGGAAATCTTCAACAGGGTCACCAACCAGCCCAATCAATGCGGAGAAACTAGAGTCTGAAGACGATGTCTCCCAAGCTTTCCTGGAGGCTGTTGCTGAGAAAAAGCCCCATGTGAAACCTTATTTCTCTAAGACCATCCGTGATCTGGAAGTCGTGGAGGGAAGCGCTGCTAGATTTGACTGCAAGATTGAAGGATACCCAGACCCTGAGGTGGTCTGGTTCAAGGATGACCAATCAATCAGGG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Martinsen et al.
Pflugers Archiv : European journal of physiology, 466(7), 1377-1389 (2013-10-29)
The Ca(2+)-dependent kinase myosin light chain kinase (MLCK) is the activator of smooth muscle contraction. In addition, it has been reported to be involved in Ca(2+) channel regulation in cultured cells, and we previously showed that the MLCK inhibitor ML-7

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service