Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

E7408

Sigma-Aldrich

p3XFLAG-CMV-7 Expression Vector

shuttle vector for transient expression of N-terminal Met-3xFLAG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

tag

Met-3X FLAG tagged

grade

for molecular biology

form

buffered aqueous solution

Peptide tag location

N-terminal

shipped in

dry ice

storage temp.

−20°C

General description

p3XFLAG-CMV-7 Expression Vector is a 4.7 kb derivative of pCMV5 used to establish expression of N-terminal Met-FLAG® fusion proteins in mammalian cells. The vector encodes the ANTI-FLAG M2 antibody epitope (Asp-Tyr-Lys-Xaa-Xaa-Asp) three times, upstream of the multiple cloning region. This results in increased detection sensitivity using ANTI-FLAG M2. The third epitope includes the enterokinase recognition sequence allowing cleavage of the 3XFLAG peptide from the purified fusion protein. Efficiency of replication is optimal when using an SV40 T antigen-expressing host.

p3XFLAG-CMV-7-BAP Control Plasmid is a 6.2 kb derivative of pCMV5 used for transient intracellular expression of N-terminal 3XFLAG bacterial alkaline phosphatase fusion protein in mammalian cells. The vector encodes three adjacent FLAG epitopes (Asp- Tyr-Lys-Xaa-Xaa-Asp) upstream of the multiple cloning region. This results in increased detection sensitivity using ANTI-FLAG M2 antibody.3 The third FLAG epitope includes the enterokinase recognition sequence, allowing cleavage of the 3XFLAG peptide from the purified fusion protein.

Vector Maps and Sequences

Components

  • p3XFLAG-CMV-7 Expression Vector 20 μg (E2400) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.
  • p3XFLAG-CMV-7-BAP Control Plasmid 20 μg (C7472) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.

Principle

The promoter-regulatory region of the human cytomegalovirus drives transcription of FLAG®-fusion constructs. p3XFLAG-CMV-7 Expression Vector is a shuttle vector for E. coli and mammalian cells.

Legal Information

This product is covered by the following patents owned by Sigma-Aldrich Co. LLC: US6,379,903, US7,094,548, JP4405125,EP1220933, CA2386471 and AU774216.
3xFLAG is a trademark of Sigma-Aldrich Co. LLC
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
p3xFLAG-CMV is a trademark of Sigma-Aldrich Co. LLC

related product

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pou5f1 contributes to dorsoventral patterning by positive regulation of vox and modulation of fgf8a expression.
Belting, H.-G., et al.
Developmental Biology, 256, 323-323 (2011)
Zaruhi Karabekian et al.
Journal of virology, 79(6), 3525-3535 (2005-02-26)
The murine cytomegalovirus (MCMV) proteins encoded by US22 genes M139, M140, and M141 function, at least in part, to regulate replication of this virus in macrophages. Mutant MCMV having one or more of these genes deleted replicates poorly in macrophages
Tatsuo Kido et al.
Journal of genetics and genomics = Yi chuan xue bao, 38(5), 181-191 (2011-05-31)
The testis specific protein Y-encoded (TSPY) is a member of TSPY/SET/NAP1 superfamily, encoded within the gonadoblastoma locus on the Y chromosome. TSPY shares a highly conserved SET/NAP-domain responsible for protein--protein interaction among TSPY/SET/NAP1 proteins. Accumulating data, so far, support the
Hyun Ju Oh et al.
Biology of reproduction, 72(2), 407-415 (2004-10-08)
In mammals, the SRY/Sry gene on the Y chromosome is necessary and sufficient for a bipotential gonad to develop into a testis, regardless of its chromosomal sex. The SRY/Sry gene encodes a protein that belongs to a high-mobility-group (HMG) box
Qi-Heng Yang et al.
Genes & development, 17(12), 1487-1496 (2003-06-20)
Omi/HtrA2 is a mitochondrial serine protease that is released into the cytosol during apoptosis to antagonize inhibitors of apoptosis (IAPs) and contribute to caspase-independent cell death. Here, we demonstrate that Omi/HtrA2 directly cleaves various IAPs in vitro, and the cleavage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service