Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HF1-5

94101909, rat liver, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

HF1-5, 94101909

biological source

rat liver

growth mode

Adherent

karyotype

100 chromosomes

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat hepatoma Hybrid [H5xFao]

Cell Line Description

HF1-5 is a subclone of HF1 selected for it′s ability to grow in glucose free medium. The entire group of hepatic functions are re-expressed. NB. The cells are difficult to cultivate, they do not like either low or high density and should be transferred frequently even though they grow slowly.

Application

Gene analysis and liver function expression.

Culture Medium

Coon′s modified Ham′s F12 (or Kaign′s modified Ham′s F12 + 45mg/L ascorbic acid + 18mg/L inositol) + 2mM Glutamine + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split semi-confluent (60%-70%) cultures 1:5 i.e. seeding at 3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 37°C. Saturation density is reached at 5x1000,000 cells/10cm dish.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service