Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Hs 633T

89050201, human fibrosarcoma, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human fibrosarcoma
growth mode:
Adherent
karyotype:
Not specified
morphology:
Not specified
products:
Not specified
receptors:
Not specified
relevant disease(s):
cancer

Product Name

Hs 633T, 89050201

biological source

human fibrosarcoma

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human fibrosarcoma MBA/8387

Cell Line Description

Human fibrosarcoma

Application

Tumourigenicity studies

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:8 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service