Recommended Products
biological source
Mouse x Mouse Hybridoma
ImmunoDon
BALB/c mouse spleen
Myeloma
NS1/1
Reactivity
Anti-CD134, rat
technique(s)
cell culture | mammalian: suitable
isotype
IgG2b
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse NS1/1 x Mouse BALB/c spleen
Cell Line Description
The antibody recognises an antigen detected only on rat T blasts that also express the CD4 antigen. The OX-40 antigen is a distinct chain of molecular weight 45-50kDa. OX-40 monoclonal antibody augments T cell proliferation at late stages of in vitro responses.
Immunogen
PHA-activated rat lymph node cells
Culture Medium
RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Seed at 5x10,000 viable cells/ml. Subculture when cells reach 5x100,000-1x1,000,000 cells/ml (usually after 3-4 days); 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service