Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B92

85042304, rat nerve, Glial/neuronal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

B92, 85042304

biological source

rat nerve

growth mode

Adherent

karyotype

Not specified

morphology

Glial/neuronal

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat nervous tissue glial

Cell Line Description

Ethyl nitrosaurea induced tumour cell line. Neuronal and glial cells are useful for research into the distribution of neurotransmitter synthesis and brain specific antigens among nerve and glia. Morphology significantly affected when grown in serum free media.

Application

Neurobiological studies

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service