Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BRIN-BG7

10020902, rat pancreas (islets), Epitheloid

Synonym(s):

BRIN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

BRIN-BG7, from rat pancreas(islets), 10020902

biological source

rat pancreas (islets)

growth mode

Adherent

karyotype

Modal chromosome number 66 up to passage 50

morphology

Epitheloid

products

BRIN-BG7 is a stable, glucose-responsive insulin secreting beta cell line expressing insulin, glucokinase and IAPP as detected by immunocytochemistry. BRIN-BG7 cells express the GLUT2 glucose transporter.

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

Cell Line Origin

A rat pancreatic Beta cell: RINm5F hybrid cell line. Insulin-secreting cell line

DNA Profile

Not specified

Culture Medium

RPMI-1640 + 2 mM Glutamine + 10% FCS / FBS

Subculture Routine

Split sub-confluent cultures (70-80%) 1:8 to 1:10 , i.e., seeding at 2-4 x 104 cells/cm2 using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37 °C. Population doubling approx 20hrs. At confluence 105 cells/cm2 can be expected.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service