Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6930

Sigma-Aldrich

Monoclonal Anti-Cytokeratin Peptide 19 antibody produced in mouse

clone A53-B/A2, tissue culture supernatant

Synonym(s):

Anti-CK19, Anti-K19, Anti-K1CS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

tissue culture supernatant

antibody product type

primary antibodies

clone

A53-B/A2, monoclonal

mol wt

antigen 40 kDa

contains

15 mM sodium azide

species reactivity

human

technique(s)

indirect immunofluorescence: 1:50 using formalin-fixed, paraffin-embedded, human tissue sections
microarray: suitable

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT19(3880)

General description

Monoclonal Anti-Cytokeratin Peptide 19 (mouse IgG2a isotype) is derived from the A53-B/A2 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with mammary human carcinoma cell line MCF-7. Monoclonal Anti-Cytokeratin Peptide 19, known as clone A53-B/A2, or clone No. Ks 19.1, reacts with the rod domain of human cytokeratin peptide 19 (40 kDa), a cytoskeletal protein restricted to epithelial and carcinoma cells. Cytokeratin 19 is a member of the type I acidic subfamily. It is differentially expressed in various human tissues.

Specificity

Also cited as clone no. KS 19.1, the antibody reacts with the rod domain of cytokeratin peptide 19, a cytoskeletal protein restricted to epithelial and carcinoma cells. Species cross-reactivity is low or absent. The antibody labels simple epithelia and basal cells of non-cornifying stratified squamous epithelia. It is a useful tool in discriminating simple carcinomas from those of different origin and for carcinoma subtyping. This antibody has also been shown to be a marker of pre-malignant lesions of the oral epithelium.

Immunogen

human mammary carcinoma cell line MCF-7.

Application

Monoclonal Anti-Cytokeratin Peptide 19 antibody produced in mouse is also suitable:
  • for immunoblotting, for immunocytochemistry,
  • for immunofluorescence,
  • as a marker of premalignant lesions of the oral epithelium, to label cytokeratin in formalin-fixed or Carnoy-fixed, paraffin embedded tissue and in frozen sections of human tissue, to label simple epithelia and basal cells of noncornifying stratified squamous epithelia.

Monoclonal Anti-Cytokeratin Peptide 19 antibody produced in mouse is suitable for microarray and indirect immunofluorescence at a dilution of 1:50 using formalin-fixed, paraffin-embedded and human tissue sections.

Biochem/physiol Actions

Keratin, type I cytoskeletal 19 (40kDa protein) is also known as cytokeratin-19 (CK-19) or keratin-19 (K19). It is encoded by the KRT19 gene in humans. Keratin 19 is a type I keratin. Keratins are intermediate filament-forming proteins that provide mechanical support and perform a variety of additional functions in epithelial cells. K19 is expressed in a subset of hepatocellular carcinomas (HCC) with poor prognosis and is considered as a biliary/hepatic progenitor cell (HPC) marker. It is also a predictor of poorer prognosis for patients with human lung squamous cell carcinoma (SCC).
Monoclonal anti-cytokeratins are specific markers of epithelial cell differentiation and have been widely used as tools in tumor identification and classification.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kasper, M., et al.
European Journal of Cancer, 23, 137-137 (1987)
Izabela Podgorski et al.
The American journal of pathology, 175(3), 1255-1269 (2009-08-25)
Bone metastasis is a hallmark of advanced prostate and breast cancers, yet the critical factors behind attraction of tumors to the skeleton have not been validated. Here, we investigated the involvement of cathepsin K in the progression of prostate tumors
Minati Satpathy et al.
Theranostics, 9(3), 778-795 (2019-02-28)
Cancer heterogeneity and drug resistance limit the efficacy of cancer therapy. To address this issue, we have developed an integrated treatment protocol for effective treatment of heterogeneous ovarian cancer. Methods: An amphiphilic polymer coated magnetic iron oxide nanoparticle was conjugated
Targeted drug delivery and image-guided therapy of heterogeneous ovarian cancer using HER2-targeted theranostic nanoparticles
Satpathy M, et al.
Theranostics, 9(3), 778-778 (2019)
R Moll et al.
Laboratory investigation; a journal of technical methods and pathology, 65(1), 74-86 (1991-07-01)
The expression of intermediate filament proteins, particularly individual cytokeratins (CKs), vimentin, and glial filament protein, was immunohistochemically investigated using frozen sections and Carnoy-fixed, paraffin-embedded tissue from normal fetal and adult human kidneys as well as from pathologically altered kidneys. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service