Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYP96450RB

Round bottom assay plate

size 96 wells, round bottom blue polypropylene wells, maximum volume 500 μL, pkg of 10x5plates/cs

Synonym(s):

96 multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
round bottom blue polypropylene wells
maximum volume:
500 μL
size:
96 wells
working volume:
4.6 mL

material

round bottom blue polypropylene wells

sterility

non-sterile

manufacturer/tradename

Axygen Inc P-96-450R-B
Corning

packaging

pkg of 10x5plates/cs

maximum volume

500 μL

size

96 wells

working volume

4.6 mL

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service