Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV50179

Sigma-Aldrich

Anti-KCNK4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-K2p4.1, Anti-Potassium channel, subfamily K, member 4, Anti-TRAAK, Anti-TRAAK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
guinea pig, human, rat
technique(s):
western blot: suitable
citations:
1

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

43 kDa

species reactivity

guinea pig, human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... KCNK4(50801)

Immunogen

Synthetic peptide directed towards the N terminal region of human KCNK4

Application

Anti-KCNK4 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

Potassium channel, subfamily K, member 4 (KCNK4) is a potassium channel protein containing two pore-forming P domains, expressed primarily in neural tissues. It has regulatory role in muscle contraction, osmotic regulation and action potential.

Sequence

Synthetic peptide located within the following region: ELGEVREKFLRAHPCVSDQELGLLIKEVADALGGGADPETNSTSNSSHSA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assignment of mouse cardiac two-pore background K+ channel gene (Kcnk4) to the proximal region of mouse chromosome 5.
A Fujita et al.
Genomics, 54(1), 183-184 (1998-11-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service