Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV49534

Sigma-Aldrich

Anti-JPH3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CAGL237, Anti-FLJ44707, Anti-HDL2, Anti-JP-3, Anti-JP3, Anti-Junctophilin 3, Anti-TNRC22

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

81 kDa

species reactivity

human, mouse, pig, bovine, dog, horse, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... JPH3(57338)

Immunogen

Synthetic peptide directed towards the N terminal region of human JPH3

Application

Anti-JPH3 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

Junctophilins are localized to the endoplasmic/sarcoplasmic reticulum membrane and anchor it to the plasma membrane. Mutations in Junctophilin 3 (JPH3) induces polyglutamine protein toxicity in Huntington′s disease.

Sequence

Synthetic peptide located within the following region: SSGGRFNFDDGGSYCGGWEDGKAHGHGVCTGPKGQGEYTGSWSHGFEVLG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian Wilburn et al.
Neuron, 70(3), 427-440 (2011-05-11)
Huntington's disease-like-2 (HDL2) is a phenocopy of Huntington's disease caused by CTG/CAG repeat expansion at the Junctophilin-3 (JPH3) locus. The mechanisms underlying HDL2 pathogenesis remain unclear. Here we developed a BAC transgenic mouse model of HDL2 (BAC-HDL2) that exhibits progressive
Lucia Golini et al.
The Journal of biological chemistry, 286(51), 43717-43725 (2011-10-25)
Junctophilins (JPs) anchor the endo/sarcoplasmic reticulum to the plasma membrane, thus contributing to the assembly of junctional membrane complexes in striated muscles and neurons. Recent studies have shown that JPs may be also involved in regulating Ca2+ homeostasis. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service