Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV44147

Sigma-Aldrich

Anti-SLC9A9 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ35613, Anti-NHE9, Anti-Nbla00118, Anti-Solute carrier family 9 (sodium/hydrogen exchanger), member 9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

72 kDa

species reactivity

mouse, horse, human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC9A9(285195)

General description

Solute carrier family 9 (sodium/hydrogen exchanger), member 9 (SLC9A9, NHE9, Nbla00118) is a member of the sodium/hydrogen exchanger family is a membrane protein that regulates the luminal pH of the recycling endosome, an essential organelle for synaptic transmission and plasticity. Disruption of SLC9A9 may contribute to the attention deficit hyperactivity disorder-like phenotype.

Specificity

Anti-SLC9A9 polyclonal antibody reacts with chicken, bovine, human, and mouse solute carrier family 9 (sodium/hydrogen exchanger), member 9 proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of human SLC9A9

Application

Anti-SLC9A9 polyclonal antibody is used to tag solute carrier family 9 (sodium/hydrogen exchanger), member 9 for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of solute carrier family 9 (sodium/hydrogen exchanger), member 9 in the regulation of luminal pH of recycling endosomes and attention deficit hyperactivity disorder-like phenotype.

Biochem/physiol Actions

SLC9A9 may act in electroneutral exchange of protons for Na(+) across membranes.

Sequence

Synthetic peptide located within the following region: INYQEQASSPCSPPARLGLDQKASPQTPGKENIYEGDLGLGGYELKLEQT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service