Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV40487

Sigma-Aldrich

Anti-EIF4G3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Eukaryotic translation initiation factor 4 γ, 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

57 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EIF4G3(8672)

General description

Eukaryotic translation initiation factor 4 gamma 3 (EIF4G3) is involved in protein translation within the mammalian mRNA ribosome complex. EIF4G3 has been shown to be a target of cytotoxic T lymphocyte (CTL) produced granzyme B (GrB) suggesting that EIF4G3 may be targeted by CTL cells as a mechanism to inhibit the use of host translational machinery by infectious viruses.

Specificity

Anti-EIF4G3 polyclonal antibody reacts with human, mouse, rat, and bovine eukaryotic translation initiation factor 4 gamma 3 proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human EIF4G3

Application

Anti-EIF4G3 polyclonal antibody is used to tag eukaryotic translation initiation factor 4 gamma 3 proteins for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the role of eukaryotic translation initiation factor 4 gamma 3 in the initiation of mRNA translation and as a target of CTL cells during infectious virus assault.

Biochem/physiol Actions

EIF4G3 is probable component of the protein complex eIF4F, which is involved in the recognition of the mRNA cap, ATP-dependent unwinding of 5′-terminal secondary structure and recruitment of mRNA to the ribosome. It is thought to be a functional homolog of EIF4G1.

Sequence

Synthetic peptide located within the following region: TAASDQKQEEKPKPDPVLKSPSPVLRLVLSGEKKEQEGQTSETTAIVSIA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Huang et al.
Journal of oncology, 2021, 4936207-4936207 (2021-07-02)
As a selective histone deacetylase (HDAC) inhibitor developed in China, chidamide has been applied for the treatment of refractory peripheral T-cell lymphoma (PTCL) and multiple solid tumors, including lung cancer. However, the underlying mechanisms are not well elucidated. In our

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service