Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV35236

Sigma-Aldrich

Anti-CACNG4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Calcium channel, voltage-dependent, γ subunit 4, Anti-MGC11138, Anti-MGC24983

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
human, rat, bovine, guinea pig, mouse, horse, rabbit
technique(s):
western blot: suitable
citations:
1

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36 kDa

species reactivity

human, rat, bovine, guinea pig, mouse, horse, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CACNG4(27092)

General description

CACNG4 is a type I transmembrane AMPA receptor regulatory protein (TARP). Studies have reported that targeted disruption in CACNG4 gene exaggerates spike-wave seizures in stargazer mouse.
Rabbit Anti-CACNG4 antibody recognizes chicken, zebrafish, bovine, human, mouse, and rat CACNG4.

Immunogen

Synthetic peptide directed towards the N terminal region of human CACNG4

Application

Rabbit Anti-CACNG4 antibody is suitable for western blot applications at a concentration of 1 μg/ml.

Biochem/physiol Actions

L-type calcium channels are composed of five subunits. CACNG4 represents one of these subunits, gamma, and is one of several gamma subunit proteins. It is an integral membrane protein that is thought to stabilize the calcium channel in an inactive (closed) state. This gene is a member of the neuronal calcium channel gamma subunit gene subfamily of the PMP-22/EMP/MP20 family.L-type calcium channels are composed of five subunits. The protein encoded by this gene represents one of these subunits, gamma, and is one of several gamma subunit proteins. It is an integral membrane protein that is thought to stabilize the calcium channel in an inactive (closed) state. This gene is a member of the neuronal calcium channel gamma subunit gene subfamily of the PMP-22/EMP/MP20 family and is located in a cluster with two similar gamma subunit-encoding genes.L-type calcium channels are composed of five subunits. The protein encoded by this gene represents one of these subunits, gamma, and is one of several gamma subunit proteins. It is an integral membrane protein that is thought to stabilize the calcium channel in an inactive (closed) state. This gene is a member of the neuronal calcium channel gamma subunit gene subfamily of the PMP-22/EMP/MP20 family and is located in a cluster with two similar gamma subunit-encoding genes.

Sequence

Synthetic peptide located within the following region: GPPPRRARGDLTHSGLWRVCCIEGIYKGHCFRINHFPEDNDYDHDSSEYL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Verity A Letts et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(6), 2123-2128 (2005-01-29)
The voltage-dependent calcium channel gamma4 subunit protein, CACNG4, is closely related to the gamma2 subunit, CACNG2. Both are expressed primarily in the brain and share 53% amino acid identity. The Cacng2 gene is disrupted in the stargazer mouse, with its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service