Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV32135

Sigma-Aldrich

Anti-MyF6 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MRF4, Anti-Myogenic factor 6 (herculin)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
polyclonal
application:
IHC
WB
species reactivity:
mouse, guinea pig, sheep, rabbit, horse, rat, goat, bovine, dog, human
technique(s):
immunohistochemistry: suitable
western blot: suitable
citations:
1

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

27 kDa

species reactivity

mouse, guinea pig, sheep, rabbit, horse, rat, goat, bovine, dog, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MYF6(4618)

General description

MyF6 is a possible helix-loop-helix protein that may be involved in muscle differentiation. It is known to have conserved DNA binding and dimerization domains.
Rabbit Anti-MyF6 antibody recognizes canine, human, mouse, rat, bovine, zebrafish, pig, and chicken MyF6.

Immunogen

Synthetic peptide directed towards the N terminal region of human MYF6

Application

Rabbit Anti-MyF6 antibody can be used for western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

MYF6 is a part of the myogenic basic helix-loop-helix family of transcription factors, and can activate the muscle differentiation program.

Sequence

Synthetic peptide located within the following region: PGLQPPHCPGQCLIWACKTCKRKSAPTDRRKAATLRERRRLKKINEAFEA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Braun et al.
Nucleic acids research, 19(20), 5645-5651 (1991-10-25)
The muscle regulatory proteins Myf3, Myf4, Myf5, and Myf6 share a highly conserved DNA binding and dimerization domain consisting of a cluster of basic amino acids and a potential helix-loop-helix structure. Here we demonstrate that the four human muscle-specific HLH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service