Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

A5581

Sigma-Aldrich

3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene

≥95%

Synonym(s):

2,2′-(Hydroxynitrosohydrazino)bis-ethanamine, DETA NONOate, NOC-18

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H13N5O2
CAS Number:
Molecular Weight:
163.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95%

form

powder

solubility

0.1 M NaOH: 10 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

NCCN(CCN)\[N+]([O-])=N/O

InChI

1S/C4H13N5O2/c5-1-3-8(4-2-6)9(11)7-10/h10H,1-6H2/b9-7+

InChI key

YNRCBOXEDICOIX-VQHVLOKHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene has been used as a nitric oxide donor supplement for neuronal culture. It has also been used as a nitric oxide donor to study the effects of protein S-nitrosation (SNO) after nitrite treatment.
3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene has been used to provide insight into the molecular pathways that may contribute to the beneficial effects of nicorandil in the cardiovascular system. 3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene causes the release of nitric oxide spontaneously in a rate-controlled manner.

Biochem/physiol Actions

3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene is a long-acting nitric oxide donor. It might exhibit nociceptive effects in rats.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shumin Tan et al.
PLoS pathogens, 9(4), e1003282-e1003282 (2013-04-18)
The ability of Mycobacterium tuberculosis (Mtb) to thrive in its phagosomal niche is critical for its establishment of a chronic infection. This requires that Mtb senses and responds to intraphagosomal signals such as pH. We hypothesized that Mtb would respond
Iris C Vermeulen Windsant et al.
Critical care (London, England), 16(3), R95-R95 (2012-05-26)
The increasing number of reports on the relation between transfusion of stored red blood cells (RBCs) and adverse patient outcome has sparked an intense debate on the benefits and risks of blood transfusions. Meanwhile, the pathophysiological mechanisms underlying this postulated
Natalia Rios et al.
Free radical biology & medicine, 101, 284-295 (2016-09-20)
The specific and sensitive detection of peroxynitrite (ONOO-/ONOOH) in biological systems is a great challenge due to its high reactivity towards several biomolecules. Herein, we validated the advantages of using fluorescein-boronate (Fl-B) as a highly sensitive fluorescent probe for the
Keigo Ueno et al.
PloS one, 6(9), e24759-e24759 (2011-09-21)
The intestinal resident Candida glabrata opportunistically infects humans. However few genetic factors for adaptation in the intestine are identified in this fungus. Here we describe the C. glabrata CYB2 gene encoding lactate dehydrogenase as an adaptation factor for survival in
A new nitric oxide donor, NOC-18, exhibits a nociceptive effect in the rat formalin model
Shibuta S, et al.
Journal of the Neurological Sciences, 141(1), 1-5 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service