Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4336

Sigma-Aldrich

4-Aminobenzoyl-Gly-Pro-D-Leu-D-Ala hydroxamic acid

≥95% (HPLC)

Synonym(s):

FN-439

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H34N6O6
CAS Number:
Molecular Weight:
490.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Assay

≥95% (HPLC)

form

powder

solubility

H2O: 1 mg/mL

storage temp.

−20°C

SMILES string

CC(C)C[C@@H](NC(=O)[C@@H]1CCCN1C(=O)CNC(=O)c2ccc(N)cc2)C(=O)N[C@H](C)C(=O)NO

InChI

1S/C23H34N6O6/c1-13(2)11-17(22(33)26-14(3)20(31)28-35)27-23(34)18-5-4-10-29(18)19(30)12-25-21(32)15-6-8-16(24)9-7-15/h6-9,13-14,17-18,35H,4-5,10-12,24H2,1-3H3,(H,25,32)(H,26,33)(H,27,34)(H,28,31)/t14-,17-,18+/m1/s1

InChI key

WWHUHFFMOPIVKB-OLMNPRSZSA-N

Amino Acid Sequence

Abz-Gly-Pro-Leu-Ala-HA

Biochem/physiol Actions

Collagenase Inhibitor

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D R Garmer et al.
Proteins, 31(1), 42-60 (1998-04-29)
We investigated the binding properties of the metalloprotease inhibitors hydroxamate, methanethiolate, and methylphosphoramidate to a model coordination site occurring in several Zn2+ metalloproteases, including thermolysin. This was carried out using both the SIBFA (sum of interactions between fragments ab initio-computed)
H Anan et al.
Journal of endodontics, 22(12), 668-673 (1996-12-01)
To investigate the effects of a combination of an antibacterial agent (ofloxacin) and a collagenase inhibitor (FN-439) in the root canal treatment of apical periodontitis, we studied the healing process of experimentally induced periapical lesions in rats by using immunohistochemical
S Odake et al.
Chemical & pharmaceutical bulletin, 39(6), 1489-1494 (1991-06-01)
To develop a potent and specific collagenase inhibitor, a series of tetrapeptidyl hydroxamic acids were synthesized, based on the previous findings with tripeptidyl derivatives (Chem. Pharm. Bull., 38, 1007-1011, 1990). Among the series of tetrapeptidyl derivatives synthesized, R-Gly-Pro-Leu-Ala-NHOH and R-Gly-Pro-D-Leu-D-Ala-NHOH
S Odake et al.
Biochemical and biophysical research communications, 199(3), 1442-1446 (1994-03-30)
Synthetic inhibitors of interstitial collagenase, tri- and tetrapeptidyl hydroxamic acids, have been developed and tested for their inhibitory activities against human matrix metalloproteinases. A water soluble inhibitor, p-NH2-Bz-Gly-Pro-D-Leu-D-Ala-NHOH (FN-439) inhibited interstitial and granulocyte collagenases, granulocyte gelatinase and skin fibroblast stromelysin
Lina María Preciado et al.
Toxins, 12(1) (2019-12-22)
Snake bite envenoming is a public health problem that was recently included in the list of neglected tropical diseases of the World Health Organization. In the search of new therapies for the treatment of local tissue damage induced by snake

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service