Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A0729

Sigma-Aldrich

Monoclonal Anti-Activin B antibody produced in mouse

clone 146807, purified immunoglobulin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

146807, monoclonal

species reactivity

human

technique(s)

neutralization: 0.6-3.0 μg/mL using Activin B-induced hemoglobin expression in the K562 human chronic myelogenous leukemia cell line. (ND50)
western blot: 1-2 μg/mL

isotype

IgG1

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... INHBB(3625)

General description

Activins are members of the TGF-β family. Activins are dimeric proteins composed of 2 β subunits. Acitivn B contains 2 β B subunits. Activins bind to the Type II Activin receptor triggering phosphorylation of Type I Acitivn receptor (ALK4) and activation of downstream pathways such as proliferation and apoptosis.
Activins are pleotropic cytokines that belong to the TGF-β superfamily. There are three forms of activin, homodimeric activin A (βA-βA), activin B (βB-βB) and the heterodimeric activin AB (βA-βB). Activins bind to type I (ActRI) or type II (ActRII) receptor complexes. The cellular targets of Activin are Smad-2, -3 and -4 that form hetromeric complex and regulate transcription of target genes. The other pathways mediated by activins are Rho A, MAPK, p38 and JNK pathways. Activins play vital role in regulation of growth, differentiation, erythropoiesis, angiogenesis, survival or neural cells and tissue repair. Activin B is reported to promote wound healing by epithelial cell proliferation and migration via the RhoA-JNK signaling pathway. It is also vital in follicular development during the estrous cycle and normal fertility in female mammals. Various reports indicate the role of activin B in normal function of pancreatic β cells and in reducing the growth of renal cell carcinoma
Monoclonal Anti-Activin B recognizes human activin B.

Immunogen

purified, CHO cell-derived, recombinant human activin B.

Application

Anti-Activin B antibody may be used for immunoblotting at a working concentration of 1-2 μg/ml. The recommended working antibody concentration for ELISA is 0.5-1.0 μg/ml. For neutralization of human activin B expressed in K562 cell line, a concentration of 0.6-3.0 μg/ml is recommended.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 5% trehalose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activin Signaling and Its Role in Regulation of Cell Proliferation, Apoptosis, and Carcinogenesis.
Chen, Y.
The Journal of Experimental Biology, 231, 534-544 (2006)
J M Young et al.
Reproduction (Cambridge, England), 143(5), 637-650 (2012-03-28)
Little is known about the role of activin B during folliculogenesis. This study investigated the expression levels of activin/inhibin subunits (βA, βB, and α), steroid enzyme, and gonadotrophin receptors in theca (TC) and granulosa cells (GC) by QPCR and activin
Yin Xia et al.
The Journal of endocrinology, 202(1), 1-12 (2009-03-11)
Activin was discovered in the 1980s as a gonadal protein that stimulated FSH release from pituitary gonadotropes and was thought of as a reproductive hormone. In the ensuing decades, many additional activities of activin were described and it was found
Ingrid Wacker et al.
Molecular and cellular biology, 29(7), 1707-1718 (2009-01-23)
The von Hippel-Lindau tumor suppressor gene (VHL) is mutated in clear cell renal cell carcinomas (RCC), leading to the activation of hypoxia-inducible factor (HIF)-mediated gene transcription. Several VHL/HIF targets, such as glycolysis, angiogenesis, cell growth, and chemotaxis of tumor cells
Philippe Bertolino et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(20), 7246-7251 (2008-05-16)
All major cell types in pancreatic islets express the transforming growth factor (TGF)-beta superfamily receptor ALK7, but the physiological function of this receptor has been unknown. Mutant mice lacking ALK7 showed normal pancreas organogenesis but developed an age-dependent syndrome involving

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service