Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91252

Sigma-Aldrich

2-Deoxyribose 5-phosphate Aldolase, E. coli K12, recombinant K12 from E. coli

≥2 U/mg

Synonym(s):

DERA, Deoxyriboaldolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
4.1.2.4.
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

form

crystals
powder

specific activity

≥2 U/mg

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

Deoxyriboaldolase catalyzes the condensation of acetaldehyde and glyceraldehyde 3-phosphate to 2-deoxyribose 5-phosphate.

Unit Definition

1 U corresponds to the amount of enzyme which converts 1 μmol 2-Deoxy-D-ribose-5-phosphate per minute at pH 7.5 and 25°C

Other Notes

The enzyme catalyzes the reversible cleavage of 2-deoxyribose-5-phosphate to acetaldehyde and D-glyceraldehyde-3-phosphate

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akio Ueno et al.
Molecular and biochemical parasitology, 173(1), 39-42 (2010-05-04)
The Toxoplasma gondii deoxyribose phosphate aldolase-like (TgDPA) gene is expressed predominantly in bradyzoites. This finding allowed us to infer that TgDPA is important in the tachyzoite-to-bradyzoite development or maintenance of cyst structure although the function of this gene is still
Akio Ueno et al.
Experimental parasitology, 121(1), 55-63 (2008-10-28)
Toxoplasma gondii undergoes stage conversion from tachyzoites to bradyzoites in intermediate hosts. There have been many reports on bradyzoite-specific genes which are thought to be involved in stage conversion. Here, we described a novel T. gondii deoxyribose phosphate aldolase-like gene
Yong-Mo Kim et al.
Journal of microbiology and biotechnology, 20(6), 995-1000 (2010-07-14)
A novel deoC gene was identified from Paenibacillus sp. EA001 isolated from soil. The gene had an open reading frame (ORF) of 663 base pairs encoding 220 amino acids with a molecular mass of 24.5 kDa. The amino acid sequence
Gerard M Lacourciere et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(14), 9150-9153 (2002-06-27)
Selenium can be metabolized for protein synthesis by two major pathways in vivo. In a specific pathway it can be inserted into polypeptide chains as the amino acid selenocysteine, as directed by the UGA codon. Alternatively, selenium can be substituted
Nobuyuki Horinouchi et al.
Bioscience, biotechnology, and biochemistry, 70(6), 1371-1378 (2006-06-24)
2-Deoxyribose 5-phosphate production through coupling of the alcoholic fermentation system of baker's yeast and deoxyriboaldolase-expressing Escherichia coli was investigated. In this process, baker's yeast generates fructose 1,6-diphosphate from glucose and inorganic phosphate, and then the E. coli convert the fructose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service