Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

84885

Sigma-Aldrich

pI-Select

set of 10 acrylamido buffer solutions (10 ml each) pK 1.0-12

Synonym(s):

Acrylamido buffers, set

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41105319

Quality Level

form

liquid

shelf life

limited shelf life

pKa 

1.0
10.3
≥12.0
3.1
3.6
4.6
6.2
7.0
8.5
9.3

storage temp.

2-8°C

Other Notes

Focusing of alkaline proteases (subtilisins) in pH 10-12 immobilized gradients

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuai Wu et al.
Journal of cellular physiology, 234(10), 19059-19072 (2019-04-18)
Retinoblastoma (RB) represents an aggressive malignancy in the eye during the period of infancy and childhood. We delineated the ability of microRNA-186 (miR-186) to influence viability, invasion, migration, angiogenesis, and apoptosis of RB via the Hedgehog signaling pathway by targeting
Can Zhao et al.
Cellular signalling, 62, 109325-109325 (2019-05-28)
Myocardial ischemia-reperfusion injury (MIRI) is recognized as a major cause of morbidity and mortality which is commonly associated with coronary artery disease. In recent studies, annexin A1 gene (ANXA1) has been discovered to be involved in the treatment for MIRI.
A Bossi et al.
Electrophoresis, 15(12), 1535-1540 (1994-12-01)
Isoelectric focusing in very alkaline immobilized pH gradients (IPG) was adopted for checking the purity and assessing the pI value of two strongly alkaline proteases: Savinase and Durazym. The first enzyme (known to be the most alkaline) contains 5 Asp
Li Liu et al.
American journal of physiology. Cell physiology, 317(2), C189-C199 (2019-05-03)
Sepsis is a systemic inflammatory response syndrome resulting from infection. This study aimed at exploring the role of microRNA-140 (miR-140) in septic mice. Wnt family member 11 (WNT11) was verified to be a target gene of miR-140 after bioinformatic prediction
Shuxian Ma et al.
Frontiers in physiology, 11, 551318-551318 (2020-11-17)
Myocardial ischemia-reperfusion (MI/R) injury is characterized by iron deposition and reactive oxygen species production, which can induce ferroptosis. Ferroptosis has also been proposed to promote cardiomyocyte death. The current study sought to define the mechanism governing cardiomyocyte death in MI/R

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service