Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

404S

Sigma-Aldrich

C4d Positive Control Slides

suitable for immunohistochemistry (formalin-fixed, paraffin-embedded sections)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

packaging

pkg of 5 slides

Quality Level

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

shipped in

ambient

storage temp.

no temp limit

General description

C4d positive control slides are intended for use as positive controls for immunohistochemical (IHC) staining using an anti-C4d antibody.

Physical form

Paraffin embedded tissue section mounted on microscope slide

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Control slide products are manufactured from human biological samples (HBS). Purchaser shall not make any attempt to trace or identify donors from whom the HBS material was obtained. Purchaser shall use any data (including genetic information), generated with or by use of the HBS only in compliance with obligations of confidentiality and non-use equivalent with all applicable international and national legislation and guidelines on the protection of personal data and confidentiality. In addition to the use restrictions contained on the product label and/or product datasheet or other applicable use terms and conditions of sale, Purchaser agrees to use the HBS solely in accordance with the World Medical Association (WMA) Declaration of Helsinki adopted in 1964 as subsequently amended by WMA on Ethical Principles for Medical Research Involving Human Subjects.

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

antibody

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service