Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

49593

Sigma-Aldrich

Celite® 110

filter aid, flux calcinated

Synonym(s):

Diatomaceous earth, flux-calcined

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
11101527
PubChem Substance ID:

quality

flux calcinated

composition

Al2O3, 0.9%
CaO, 3.7%
Fe2O3, 0.4%
K2O, 0.2%
MgO, 0.3%
Na2O, 0.9%
SiO2, 93.5%

loss

≤0.1% loss on ignition, 900°C

median particle size

≤45 μm

pH

~10.5 (25 °C, 10% in slurry)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimum immobilization of Candida rugosa lipase on Celite by RSM.
Chang SF, et al.
Applied Clay Science, 37(1), 67-73 (2007)
Wenhui Xiong et al.
Journal of colloid and interface science, 368(1), 528-532 (2011-12-14)
This paper presents a novel technique integrating bulk-sensitive and surface-sensitive XANES methods to distinguish between physisorption and chemisorption for phosphate adsorption onto ferrihydrite-modified diatomite (FHMD). XANES P K-edge, L-edge, and Fe M-edge spectra were obtained for reference samples (K(2)HPO(4) and
Xiao-Li Yang et al.
Bioresource technology, 101(23), 9178-9184 (2010-08-06)
This paper examined the effect of diatomite addition on membrane fouling and process performance in an anoxic/oxic submerged membrane bioreactor (A/O MBR). Particle size distribution, molecular weight distribution and microbial activity have been investigated to characterize the sludge mixed liquor.
Necla Calışkan et al.
Combinatorial chemistry & high throughput screening, 13(8), 703-711 (2010-04-30)
This paper is the first report describing the characterization of local diatomite of Caldiran-Van region (Eastern Anatolia, Turkey). Special attention was paid to the ability of its electroanalytical performance at modified electrodes and to the potential application of diatomite-modified electrode.
Gang Pan et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 13(8), 2205-2212 (2012-01-25)
The formation mechanism of a nanoscale gas state is studied on inorganic clay surfaces modified with hexamethyldisilazane, which show different contact angles in ethanol-water solutions. As the dissolved oxygen becomes oversaturated due to the decrease in ethanol-water ratio, oxygen nanoscale

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service