Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

25023

Sigma-Aldrich

Sodium acetate

puriss., anhydrous, ≥98%, powder

Synonym(s):

Acetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa
CAS Number:
Molecular Weight:
82.03
Beilstein:
3595639
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

grade

puriss.

Assay

≥98%

form

powder
solid

autoignition temp.

1112 °F

impurities

≤0.002% heavy metals (as Pb)

loss

≤1% loss on drying, 120 °C

pH

7.5-9.2 (20 °C, 5%)

pKa 

4.76 (acetic acid)

mp

>300 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Fe: ≤10 mg/kg

SMILES string

[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 995-995 (1994)
The liquid-chromatographic quantification of some synthetic colorants in soft drinks.
Trandafir I, et al.
Scientific Study and Research, 10(1), 73-82 (2009)
R E Kingston et al.
Current protocols in molecular biology, Chapter 4, Unit4-Unit4 (2008-02-12)
Three different methods for RNA preparation using guanidine are presented in this unit--a single-step isolation method employing liquid-phase separation to selectively extract total RNA from tissues and cultured cells and two methods that rely on a CsCl step gradient to
Listeria monocytogenes inhibition in refrigerated vacuum packaged turkey bologna by chemical additives.
Wederquist HJ, et al.
Journal of Food Science, 59(3), 498-500 (1994)
Carol L Ladner-Keay et al.
PloS one, 9(6), e98753-e98753 (2014-06-04)
The formation of β-sheet rich prion oligomers and fibrils from native prion protein (PrP) is thought to be a key step in the development of prion diseases. Many methods are available to convert recombinant prion protein into β-sheet rich fibrils

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service