Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

98956

Supelco

N-Cinnamoylglycine

analytical standard

Synonym(s):

N-(1-Oxo-3-phenyl-2-propen-1-yl)glycine, Cinnamoyl glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO3
CAS Number:
Molecular Weight:
205.21
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

InChI

1S/C11H11NO3/c13-10(12-8-11(14)15)7-6-9-4-2-1-3-5-9/h1-7H,8H2,(H,12,13)(H,14,15)/b7-6+

InChI key

YAADMLWHGMUGQL-VOTSOKGWSA-N

Biochem/physiol Actions

Cinnamoylglycine is known as a urinary metabolite in man, although whether it is formed de novo from plant cinnamate or is a plant product excreted unchanged has not been conclusively demonstrated. When cinnamoylglycine occurs naturally it is probably a food constituent excreted unchanged. It is not found when small quantities (0.5-6 g) of cinnamic acid are fed to man, but by analogy with animal experiments may be produced when much larger quantities are given.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G K Brown et al.
Journal of chromatography, 145(2), 177-184 (1978-03-01)
The profile of high boiling point organic acids in urine samples from both normal subjects and patients suspected of having some form of metabolic disorder has been determined by combined gas chromatography-mass spectrometry. Fifteen different compounds eluting after hippuric acid
J A Hoskins et al.
Biomedical mass spectrometry, 11(6), 296-300 (1984-06-01)
The enzyme phenylalanine ammonia lyase taken orally has been found to reduce the rise in blood phenylalanine that normally occurs following a protein meal. Therefore the enzyme has a potential use in the management of the genetic disease phenylketonuria. The
Bejan J Saeedi et al.
Cell metabolism, 31(5), 956-968 (2020-03-28)
Many studies have suggested a role for gut-resident microbes (the "gut microbiome") in modulating host health; however, the mechanisms by which they impact systemic physiology remain largely unknown. In this study, metabolomic and transcriptional profiling of germ-free and conventionalized mouse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service