Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

94876

Supelco

Sulbactam

analytical standard

Synonym(s):

(2S,5R)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-dioxide, Betamaze, CP 45899, CP-45,899, Penicillanic acid 1,1-dioxide, Penicillanic acid S,S-dioxide, Penicillanic acid dioxide, Penicillanic acid sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO5S
CAS Number:
Molecular Weight:
233.24
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O

InChI

1S/C8H11NO5S/c1-8(2)6(7(11)12)9-4(10)3-5(9)15(8,13)14/h5-6H,3H2,1-2H3,(H,11,12)/t5-,6+/m1/s1

InChI key

FKENQMMABCRJMK-RITPCOANSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shu Xia et al.
PloS one, 9(7), e100707-e100707 (2014-07-02)
To investigate CTX-M genotypes among extended-spectrum β-lactamase-producing Escherichia coli (ESBL-EC) isolated from patients with community-onset and hospital-onset infections in China, their clonality and the distribution of CTX-M variants in different specimens of community-onset and hospital-onset infections. ESBL-EC isolates were collected
A S Levin
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases, 8(3), 144-153 (2002-05-16)
Recent studies have highlighted the emergence of infections involving multiresistant Acinetobacter clinical isolates. Sulbactam offers direct antimicrobial activity against Acinetobacter species. Accordingly, co-administration of sulbactam with ampicillin or cefoperazone offers the potential of effective empirical therapy against Acinetobacter and other
M Akova
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases, 14 Suppl 1, 185-188 (2007-12-25)
Sulbactam irreversibly inhibits the hydrolytic activity of beta-lactamases. This compound is commercially available in combination with either ampicillin or cefoperazone. In each instance, the activity of the partner antibiotic against beta-lactamase-producing bacteria is restored. One of the particular advantages of
J D Williams
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 24(3), 494-497 (1997-03-01)
beta-Lactamase inhibitors such as sulbactam are beta-lactam compounds that have low antimicrobial activity but are able to inhibit enzymes (beta-lactamases) that destroy beta-lactam antibiotics like penicillins and cephalosporins. The main activity of beta-lactamase inhibitors is directed against plasmid-mediated transferable enzymes
G S Singh
Mini reviews in medicinal chemistry, 4(1), 93-109 (2004-02-03)
Beta-lactam ring-containing compounds such as penicillins, ampicillin, amoxicillin, cephalosporins and carbapenems are among the most famous antibiotics. This article reviews the recent developments in study of cephems, oxacephems, penams and sulbactam. Many of the compounds reviewed have potential antibacterial activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service