Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91519

Supelco

2-Methylbutyryl-L-carnitine

analytical standard

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-(2-methyl-1-oxobutoxy)-1-propanaminium inner salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23NO4
CAS Number:
Molecular Weight:
245.32
Beilstein:
5946881
MDL number:
UNSPSC Code:
41116107
NACRES:
NB.21

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

optical activity

[α]/D -19.0±2.0°, c = 0.1 in H2O

shelf life

limited shelf life, expiry date on the label

impurities

≤12.0% water (calculated from elemental analysis)

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

C[N+](C)(C)C[C@H](OC(C(CC)C)=O)CC([O-])=O

InChI

1S/C12H23NO4/c1-6-9(2)12(16)17-10(7-11(14)15)8-13(3,4)5/h9-10H,6-8H2,1-5H3/t9?,10-/m1/s1

InChI key

IHCPDBBYTYJYIL-QVDQXJPCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methylbutyryl-L-carnitine belongs to the acyl-L-carnitine class of compounds, which are the acetylated forms of L-carnitine, derived from the breakdown of amino and fatty acids.

Application

2-Methylbutyryl-L-carnitine may also be used as an analytical reference standard for the separation and identification of underivatized 2-methylbutyryl-L-carnitine in human plasma samples using high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minzhi Peng et al.
Journal of chromatography. A, 1319, 97-106 (2013-10-31)
A simple HPLC-MS/MS method has been established to separate and identify underivatized acylcarnitine isomers. Human plasma samples were deproteinized and concentrated. Acylcarnitines were separated on a reverse phase column and detected with triple quadrupole linear ion trap mass spectrometry. Deuterium-labeled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service