Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91224

Supelco

Ethyl phenylacetate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2COOC2H5
CAS Number:
Molecular Weight:
164.20
Beilstein:
509140
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.497 (lit.)
n20/D 1.497-1.499

bp

229 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCOC(=O)Cc1ccccc1

InChI

1S/C10H12O2/c1-2-12-10(11)8-9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

InChI key

DULCUDSUACXJJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl phenylacetate is an organic compound, which is widely used as a flavoring agent.

Application

Ethyl phenylacetate may be used as a reference standard in the determination of the analyte in soy sauce using gas chromatography coupled with mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

210.0 °F - closed cup

Flash Point(C)

98.9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Encyclopedia of Food and Color Additives (1997)
Encyclopedia of Food and Color Additives (1997)
Identification of volatile components in shoyu (soy sauce) by gas chromatography-mass spectrometry.
Nunomura N, et al.
Agricultural and Biological Chemistry, 40(3), 485-490 (1976)
Pingxi Xu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16592-16597 (2014-10-29)
Insect repellents are important prophylactic tools for travelers and populations living in endemic areas of malaria, dengue, encephalitis, and other vector-borne diseases. DEET (N,N-diethyl-3-methylbenzamide) is a 6-decade-old synthetic repellent, which is still considered the gold standard of mosquito repellents. Mosquitoes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service