Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

84086

Millipore

Sabouraud 2% Glucose Agar

suitable for microbiology

Synonym(s):

Dermasel Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171605
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 17 g/L
D(+)-glucose, 20 g/L
special peptone, 10 g/L

final pH

5.6±0.2 (25 °C)

application(s)

cleaning products
clinical testing
cosmetics
environmental
food and beverages
personal care

microbiology

suitability

selective for Aspergillus spp.
selective for Candida spp.
selective for Penicillium spp.
selective for Pichia spp.
selective for Saccharomyces spp.
selective for Zygosaccharomyces spp.
selective for molds (General Media)
selective for yeasts (General Media)

Application

For the cultivation, isolation and identification of pathogenic fungi and yeasts.

Other Notes

Thermoresistant propagules of Neosartorya fischeri; some ecological considerations.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Z. Jesenska et al.
Journal of Food Protection, 54, 582-582 (1991)
Martyna Mroczyńska et al.
Microorganisms, 8(2) (2020-02-26)
Triazoles are the only compounds used as antibiotics in both medicine and agriculture. The presence of triazoles in the environment can contribute to the acquisition of azole resistance among isolates of Aspergillus fumigatus. The objective of this study was to

Articles

Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service