Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78336

Supelco

Dichloran

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2,6-Dichloro-4-nitroaniline, Dichloran

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(NO2)NH2
CAS Number:
Molecular Weight:
207.01
Beilstein:
1459581
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

mp

190-192 °C (lit.)

format

neat

SMILES string

Nc1c(Cl)cc(cc1Cl)[N+]([O-])=O

InChI

1S/C6H4Cl2N2O2/c7-4-1-3(10(11)12)2-5(8)6(4)9/h1-2H,9H2

InChI key

BIXZHMJUSMUDOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service