Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78109

Supelco

Isoorientin

analytical standard

Synonym(s):

Homoorientin, Luteolin 6-C-β-D-glucoside, Luteolin 6-C-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% water

application(s)

food and beverages

format

neat

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)cc3OC(=CC(=O)c3c2O)c4ccc(O)c(O)c4

InChI

1S/C21H20O11/c22-6-14-17(27)19(29)20(30)21(32-14)16-11(26)5-13-15(18(16)28)10(25)4-12(31-13)7-1-2-8(23)9(24)3-7/h1-5,14,17,19-24,26-30H,6H2/t14-,17-,19+,20-,21+/m1/s1

InChI key

ODBRNZZJSYPIDI-VJXVFPJBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isoorientin belongs to the class of C-glycosylflavones and is a major constituent of Passiflora edulis fruit.

Application

Isoorientin may be used as an analytical reference standard for the quantification of the analyte in Passiflora edulis fruit pulp and Patrinia villosa Juss using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

C-glycosyl flavone with anti-inflammatory, antimicrobial and antioxidant properties. Induces antioxidant response through PI3K singaling.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of isoorientin and total flavonoids in Passiflora edulis fruit pulp by HPLC-UV/DAD
Zeraik M.L and Yariwake.H.J
Journal of Chromatography A, 96(1), 86-91 (2010)
Dong-Seon Kim et al.
Planta medica, 76(7), 678-682 (2009-12-05)
We aimed to find antiallergic agents from natural sources using mast cells activated during allergic reaction. We screened over 2000 plants for blockade of histamine release and identified two of them, S. baicalenesis and P. edulis. Bioassay-guided fractionation led to
Mala Ranghoo-Sanmukhiya et al.
Phytochemical analysis : PCA, 21(6), 566-574 (2010-09-08)
Aloe tormentorii, A. purpurea and A. macra are used as multipurpose folk medicines in Réunion and Mauritius Islands and are mistaken for the introduced Aloe vera. To compare the phytochemical, antimicrobial and DNA profiles of Aloe endemic to Mauritius and
Roxana Orrego et al.
Molecules (Basel, Switzerland), 14(10), 3906-3913 (2009-11-20)
This study assessed the inhibitory effect of three C-glycosylflavonoids from Cymbopogon citratus leaves--isoorientin (1), swertiajaponin (2) and isoorientin 2"-Orhamnoside (3)--on human LDL oxidation. Isolated LDL was incubated with compounds 1-3 and the kinetics of lipid peroxidation were assessed by conjugated
Li Yuan et al.
Toxicology and applied pharmacology, 265(1), 83-92 (2012-10-03)
Isoorientin (ISO) is a flavonoid compound that can be extracted from several plant species, such as Phyllostachys pubescens, Patrinia, and Drosophyllum lusitanicum; however, its biological activity remains poorly understood. The present study investigated the effects and putative mechanism of apoptosis

Protocols

Effective analysis of passion flower by HPTLC fingerprinting, in conjunction with a comprehensive range of related reference materials, enables fast and efficient identification of this plant material.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service