Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77089

Supelco

Notoginsenoside R1

analytical standard

Synonym(s):

(3β,6α,12β)-20-(β-D-Glucopyranosyloxy)-3,12-dihydroxydammar-24-en-6-yl 2-O-β-D-xylopyranosyl-β-D-glucopyranoside, Sanchinoside R1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H80O18
CAS Number:
Molecular Weight:
933.13
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C\C(C)=C\CC[C@](C)(O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H]2CC[C@]3(C)[C@@H]2[C@H](O)C[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5[C@H](C[C@@]34C)O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O[C@@H]7OC[C@@H](O)[C@H](O)[C@H]7O

InChI

1S/C47H80O18/c1-21(2)10-9-13-47(8,65-41-37(59)34(56)32(54)26(18-48)62-41)22-11-15-45(6)30(22)23(50)16-28-44(5)14-12-29(52)43(3,4)39(44)25(17-46(28,45)7)61-42-38(35(57)33(55)27(19-49)63-42)64-40-36(58)31(53)24(51)20-60-40/h10,22-42,48-59H,9,11-20H2,1-8H3/t22-,23+,24+,25-,26+,27+,28+,29-,30-,31-,32+,33+,34-,35-,36+,37+,38+,39-,40-,41-,42+,44+,45+,46+,47-/m0/s1

InChI key

LLPWNQMSUYAGQI-OOSPGMBYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Notoginsenoside R1 has been shown to exhibit antooxidant, anti-inflammatory, antiapoptotic, and immune-stimulatory properties.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kouichi Yoshizaki et al.
Chemical & pharmaceutical bulletin, 60(3), 354-362 (2012-03-03)
Seven new dammarane-type triterpenoid saponins, chikusetsusaponin FK1 (1), chikusetsusaponin FK2 (2), chikusetsusaponin FK3 (3), chikusetsusaponin FK4 (4), chikusetsusaponin FK5 (5), chikusetsusaponin FK6 (6), and chikusetsusaponin FK7 (7), and eleven known triterpenoid saponins, ginsenoside Rb3 (9), ginsenoside Rc (10), chikusetsusaponin VI
Wei-Xing Chen et al.
World journal of gastroenterology, 14(1), 29-37 (2008-01-08)
To assess the effect of notoginsenoside R1 on hepatic microcirculatory disturbance induced by gut ischemia/reperfusion (I/R) in mice. The superior mesenteric artery (SMA) of C57/BL mice was ligated for 15 min to induce gut ischemia followed by 30-min reperfusion. In
Hong-Sheng Zhang et al.
Free radical biology & medicine, 40(9), 1664-1674 (2006-04-25)
The matrix fibronectin protein plays an important role in vascular remodeling. Notoginsenoside R1 is the main ingredient with cardiovascular activity in Panax notoginseng; however, its molecular mechanisms are poorly understood. We report that notoginsenoside R1 significantly decreased TNF-alpha-induced activation of
T T X Dong et al.
Phytotherapy research : PTR, 19(8), 684-688 (2005-09-24)
The root of Panax notoginseng (Radix Notoginseng, Sanqi) is a commonly used traditional Chinese medicine, which is mainly cultivated in Wenshan of Yunnan China. The identified active constituents in Radix Notoginseng include saponin, ssavonoid and polysaccharide; however, the levels of
Hong-Xiang Sun et al.
Chemistry & biodiversity, 3(7), 718-726 (2006-12-29)
The further purification of the total saponins from the roots of Panax notoginseng (Burk.) F. H. Chen by ordinary and reversed-phase silica-gel, as well as Sephadex LH-20 chromatography afforded two adjuvant active dammarane-type saponins, ginsenoside Re (1) and notoginsenoside R1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service