Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68922

Supelco

Picroside II

analytical standard

Synonym(s):

(1aS,1bS,2S,5aR,6S,6aS)-1a,1b,2,5a,6,6a-Hexahydro-6-[(4-hydroxy-3-methoxybenzoyl)oxy]-1a-(hydroxymethyl)oxireno[4,5]cyclopenta[1,2-c]pyran-2-yl-β-D-glucopyranoside, 6-Vanillylcatalpol, Amphicoside II, Vanilloyl catalpol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28O13
CAS Number:
Molecular Weight:
512.46
EC Number:
UNSPSC Code:
85151701

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

form

powder

shelf life

limited shelf life, expiry date on the label

storage temp.

−20°C

SMILES string

OC1[C@@H](O)[C@H](O)C(CO)O[C@H]1O[C@@H]2OC=C[C@]3([H])[C@@]2([H])[C@](O4)(CO)[C@@H]4[C@H]3OC(C5=CC=C(O)C(OC)=C5)=O

InChI

1S/C23H28O13/c1-31-12-6-9(2-3-11(12)26)20(30)34-18-10-4-5-32-21(14(10)23(8-25)19(18)36-23)35-22-17(29)16(28)15(27)13(7-24)33-22/h2-6,10,13-19,21-22,24-29H,7-8H2,1H3

InChI key

AKNILCMFRRDTEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Phytochemical found in Traditional Chinese Medicine herbal preparations. Picroside II, a glucoside, is reported to have hepatoprotective, cardioprotective, and neuroprotective properties.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service