Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

53158

Supelco

Hexylbenzene

analytical standard

Synonym(s):

1-Phenylhexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)5CH3
CAS Number:
Molecular Weight:
162.27
Beilstein:
1904786
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.8% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.486 (lit.)
n20/D 1.486

bp

226 °C (lit.)

mp

−61 °C (lit.)

density

0.861 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CCCCCCc1ccccc1

InChI

1S/C12H18/c1-2-3-4-6-9-12-10-7-5-8-11-12/h5,7-8,10-11H,2-4,6,9H2,1H3

InChI key

LTEQMZWBSYACLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hexylbenzene has been used as an analytical standard to analyze the elution performance of the analyte in water as eluent by high performance liquid chromatography (HPLC). It may be used as a reference standard for the determination of the analyte in water by gas chromatography-mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Compatibility

This substance has been tested against several types of hand protection for CE compliance. See related products below to find the recommended gloves for handling this product.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

suggested gloves for splash protection

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improving HPLC separation performance using parallel segmented flow chromatography
Camenzuli M, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 111, 3-7 (2013)
Characterization and source identification of hydrocarbons in water samples using multiple analytical techniques
Wang Z, et al.
Journal of Chromatography A, 971(1-2), 173-184 (2002)
Hiroshi Kobayashi et al.
Journal of chromatography. A, 1109(1), 2-9 (2005-12-20)
The contribution of molecular diffusion to peak broadening was studied in a reversed-phase HPLC system, consisting of a monolithic silica C18 column and methanol-water mobile phase. Study on the band broadening effect of holding a solute in a column or
Metabolism of n-butyl, n-hexyl, and n-octyl benzene in the bile of rainbow trout.
J Hellou et al.
Bulletin of environmental contamination and toxicology, 39(2), 182-187 (1987-08-01)
Yi Min et al.
Journal of chromatography. A, 1356, 148-156 (2014-07-08)
In this work, 1.9 μm reversed-phase packing materials with superficially porous structure were prepared to achieve the rapid and high efficient separation of peptides and proteins. The silica particles were synthesized via three steps, nonporous silica particle preparation by a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service