Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46411

Supelco

Medroxyprogesterone

VETRANAL®, analytical standard

Synonym(s):

17α-Hydroxy-6α-methyl-4-pregnene-3,20-dione, 17α-Hydroxy-6α-methylprogesterone, 6α-Methyl-5-pregnen-17α-ol-3,20-dione, 6-Dihydromegestrol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32O3
CAS Number:
Molecular Weight:
344.49
Beilstein:
2510965
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12C[C@H](C)C3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(C)=O

InChI

1S/C22H32O3/c1-13-11-16-17(20(3)8-5-15(24)12-19(13)20)6-9-21(4)18(16)7-10-22(21,25)14(2)23/h12-13,16-18,25H,5-11H2,1-4H3/t13-,16+,17-,18-,20+,21-,22-/m0/s1

InChI key

FRQMUZJSZHZSGN-HBNHAYAOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Much of the pharmacology literature refers to medroxyprogesterone acetate as medroxyprogesterone, but they are of course not the same substance.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Manes et al.
Reproduction in domestic animals = Zuchthygiene, 47(3), 412-418 (2011-09-03)
Previous research from our laboratory in beef cattle suggests that the pre-ovulatory follicle size, maturity and subsequent susceptibility to gonadotropin are influenced by the length of progestagen treatment in artificial insemination programme in beef cows. To test this hypothesis, two
M F Martinez et al.
Reproduction in domestic animals = Zuchthygiene, 47(2), 230-237 (2011-09-03)
Two experiments were designed to investigate the administration of intravaginal progesterone in protocols for oestrus and ovulation synchronization in beef heifers. In Experiment 1, cyclic Black Angus heifers (n = 20) received an Ovsynch protocol and were randomly assigned to
Carolyn J Crandall et al.
Breast cancer research and treatment, 132(1), 275-285 (2011-11-02)
The associations between breast tenderness during use of conjugated equine estrogen (CEE) therapy with or without medroxyprogesterone (MPA) therapy and subsequent breast cancer risk are unknown. We analyzed data from the Women's Health Initiative Estrogen plus Progestin (N = 16,608
D R Reilly et al.
Canadian journal of psychiatry. Revue canadienne de psychiatrie, 45(6), 559-563 (2000-09-15)
To propose a protocol for minimizing medical complications associated with the use of cyproterone, medroxyprogesterone, and depot leuprolide to treat paraphilia. Review of the relevant literature. Certain patient populations should not be treated with these medications, and medical complications associated
Toshio Hayashi et al.
Nitric oxide : biology and chemistry, 24(4), 199-203 (2011-04-26)
A comparison between the atheroprotective and osteoprotective effects of the selective estrogen receptor modulator (SERM) raloxifene and those of hormone replacement therapy (HRT) has not been made in elderly women. A randomized prospective controlled trial was performed in a cohort

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service