Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46198

Supelco

(+)-Fenchol

analytical standard

Synonym(s):

(1R)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-ol, Fenchyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
2038082
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (sum of enantiomers, GC)

optical activity

[α]20/D +11.0±0.5°, c = 10% in ethanol

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

201-202 °C (lit.)

mp

43-46 °C

format

neat

SMILES string

CC1(C)[C@H]2CC[C@](C)(C2)[C@H]1O

InChI

1S/C10H18O/c1-9(2)7-4-5-10(3,6-7)8(9)11/h7-8,11H,4-6H2,1-3H3/t7-,8-,10+/m0/s1

InChI key

IAIHUHQCLTYTSF-OYNCUSHFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Chiral building block

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A.A. van der Zeijden et al.
Synthesis, 847-847 (1996)
Y. Yuasa et al.
Tetrahedron, 48, 3473-3473 (1992)
M Miyazawa et al.
Xenobiotica; the fate of foreign compounds in biological systems, 37(9), 943-953 (2007-11-10)
The metabolism of (+)-fenchol was investigated in vitro using liver microsomes of rats and humans and recombinant cytochrome P450 (P450 or CYP) enzymes in insect cells in which human/rat P450 and NADPH-P450 reductase cDNAs had been introduced. The biotransformation of
Sergio Abbate et al.
Chirality, 21 Suppl 1, E242-E252 (2009-11-21)
The first well documented experiments of Near Infrared Vibrational Circular Dichroism (NIR-VCD) were performed around 1975. We review the thirty year history of NIR-VCD, encompassing both instrumental development and theoretical/computational methods that allow interpretation of experimental spectra, harvesting useful structural
D M Satterwhite et al.
The Journal of biological chemistry, 260(26), 13901-13908 (1985-11-15)
The conversion of geranyl pyrophosphate to (-)-endo-fenchol is considered to proceed by the initial isomerization of the substrate to (-)-(3R)-linalyl pyrophosphate and the subsequent cyclization of this bound intermediate. To test this stereochemical scheme, phosphatase-free preparations of (-)-endo-fenchol cyclase from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service