Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45831

Supelco

Chlorpyrifos-methyl solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7Cl3NO3PS
CAS Number:
Molecular Weight:
322.53
Beilstein:
1541078
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

COP(=S)(OC)Oc1nc(Cl)c(Cl)cc1Cl

InChI

1S/C7H7Cl3NO3PS/c1-12-15(16,13-2)14-7-5(9)3-4(8)6(10)11-7/h3H,1-2H3

InChI key

HRBKVYFZANMGRE-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R C Malima et al.
Medical and veterinary entomology, 23(4), 317-325 (2009-11-28)
Three insecticides - the pyrethroid deltamethrin, the carbamate carbosulfan and the organophosphate chlorpyrifos-methyl - were tested on mosquito nets in experimental huts to determine their potential for introduction as malaria control measures. Their behavioural effects and efficacy were examined in
G A R Silva et al.
Bulletin of entomological research, 101(2), 177-185 (2010-11-03)
Oryzaephilus mercator and O. surinamensis are stored grains and processed food pests, the latter being responsible for major economical losses. Polyacrylamide gel electrophoresis was used to analyse esterase patterns during insect development. Seven esterases, three cholinesterases, two carboxylesterases and two
Yael Merbl et al.
Journal of zoo and wildlife medicine : official publication of the American Association of Zoo Veterinarians, 42(1), 144-148 (2011-03-01)
A 5-yr-old spayed female caracal (Caracal caracal) was presented with complaints of acute onset of mental dullness, ataxia, and ventroflexion of 24-hr duration. The animal's garden territory was sprayed a day earlier with an organophosphate (OP) insecticide (chlorpyriphos-methyl). The caracal
R Riccio et al.
Food additives and contaminants, 23(7), 683-692 (2006-06-06)
The effects of the cuticle and epicuticular waxes of grapefruit, strawberry and apple on the photodegradation and penetration of chlorpyrifos-methyl were studied. Photodegradation experiments were conducted by exposing the insecticide to the light of a xenon lamp in the presence
Bartosz Wielgomas et al.
Analytical and bioanalytical chemistry, 390(7), 1933-1941 (2008-02-27)
The present work describes a headspace single-drop microextraction (HS-SDME) method in conjunction with gas chromatography electron capture detection (GC-ECD) for the determination of an organophosphate insecticide, chlorpyrifos-ethyl (CPF), in rat liver. Sample preparation included tissue homogenization with methanol in the

Protocols

analytical standard; Tokuthion, technical grade, pkg of 50 mg; Crotoxyphos; Phorate; Azinphos-ethyl; Diazinon; Azinphos-methyl; Demeton-O; Dimethoate; Chlorpyrifos-methyl

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service