Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

45634

Supelco

Promecarb

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO2
CAS Number:
Molecular Weight:
207.27
Beilstein:
2111695
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)Oc1cc(C)cc(c1)C(C)C

InChI

1S/C12H17NO2/c1-8(2)10-5-9(3)6-11(7-10)15-12(14)13-4/h5-8H,1-4H3,(H,13,14)

InChI key

DTAPQAJKAFRNJB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Promecarb is a broad spectrum pesticide containing the N-methylcarbamate group. Its mode of action involves inhibiting the cholinesterase enzyme activity.

Application

Promecarb may be used as a reference standard for the determination of promecarb in water by solid-phase extraction and liquid chromatography coupled with ionspray mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saeid Khodadoust et al.
Analytica chimica acta, 699(1), 113-119 (2011-06-28)
A rapid and simple method for the extraction and preconcentration of N-methylcarbamates (NMCs) (carbofuran, carbaryl and promecarb) in water samples using dispersive liquid-liquid microextraction (DLLME) using chemometrics was developed. Influence variables such as volume of extracting (CHCl(3)) and dispersing solvents
F García Sánchez et al.
The Analyst, 116(8), 851-856 (1991-08-01)
The use of spectrofluorimetry to determine the fluorescent derivative of the insecticide promecarb, following hydrolysis to the corresponding phenol in basic media and subsequent coupling with the labelling agent dansyl chloride, is described and discussed. A study of media of
Tomas Pérez-Ruiz et al.
The Analyst, 127(11), 1526-1530 (2002-12-12)
A flow injection-chemiluminescent method for the determination of carbofuran and promecarb is proposed. The assay was based on the on-line conversion of the pesticides into methylamine by irradiation with UV light. The methylamine generated was subsequently reacted with tris(2,2'-bipyridine)ruthenium(III), which
Determination of poorly fluorescent carbamate pesticides in water, bendiocarb and promecarb, using cyclodextrin nanocavities and related media.
Pacioni N L and Veglia A V
Analytica Chimica Acta, 583(1), 63-71 (2007)
H F Krug et al.
The Biochemical journal, 250(1), 103-110 (1988-02-15)
Carbamates are a widely used class of insecticides and herbicides. They were tested for their ability to affect human blood platelet aggregation and arachidonic acid metabolism in platelets. (1) The herbicides of the carbamate type have no, or only little

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service