Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44006

Supelco

Avicularin

analytical standard

Synonym(s):

3,3′,4′,5,7-Pentahydroxyflavone 3-O-α-L-arabinofuranoside, Avicularoside, Quercetin 3-O-α-L-arabinofuranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18O11
CAS Number:
Molecular Weight:
434.35
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% water

application(s)

food and beverages

format

neat

SMILES string

OC1=CC(O)=CC2=C1C(C(O[C@@H]3O[C@H]([C@@H]([C@H]3O)O)CO)=C(O2)C4=CC(O)=C(O)C=C4)=O

InChI

1S/C20H18O11/c21-6-13-15(26)17(28)20(30-13)31-19-16(27)14-11(25)4-8(22)5-12(14)29-18(19)7-1-2-9(23)10(24)3-7/h1-5,13,15,17,20-26,28H,6H2/t13-,15-,17+,20-/m0/s1

InChI key

BDCDNTVZSILEOY-UXYNSRGZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Q Zhang et al.
Proceedings of the Chinese Academy of Medical Sciences and the Peking Union Medical College = Chung-kuo i hsueh k'o hsueh yuan, Chung-kuo hsieh ho i k'o ta hsueh hsueh pao, 4(4), 193-195 (1989-01-01)
A method for the determination of avicularin in Polygonum aviculare L. is described. Square wave polarography (SWP) with a static mercury drop electrode was used to observe the reduction of avicularin. The supporting electrolyte was an acetate buffer solution of
O Servettaz et al.
Journal of natural products, 47(5), 809-814 (1984-09-01)
Six flavonol glycosides and ent-epicatechin were isolated from Dryas octopetala and their structures elucidated by chemical and spectroscopic methods. Two new flavonoids, 3-O-alpha-L-arabinofuranosyl-8-methoxyquercetin (2) and 3-O-beta-galactopyranosyl-8-methoxykaempferol (6), were identified along with 3-O-beta-D-galactopyranosylquercetin (hyperin) (3), 3-O-alpha-L-arabinofuranosylquercetin (avicularin) (5), 3-O-beta-L-arabinopyranosylquercetin (4), and
Jian-Feng Xu et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 30(6), 669-671 (2007-10-09)
To investigate the chemical constituents from the branches and leaves of Cupressus duolouxiana native to China in order to obtain a more comprehensive understanding of its effective components. Compounds were isolated and purified by silica gel column chromatography. They were
Shu-Qi Wang et al.
Phytochemistry, 87, 119-125 (2013-01-02)
Three biflavonoid glycosides along with 12 known flavonoids, were isolated from leaves of Malus hupehensis. The complete structures of two of the compounds were established from analysis of MS, NMR spectroscopic and CD data, as well as DFT CD calculations
Yan Wang et al.
Journal of enzyme inhibition and medicinal chemistry, 21(1), 87-93 (2006-03-31)
The medicinal herb parasitic loranthus in a screen was found to inhibit fatty acid synthase (EC 2.3.1.85, FAS) and reduce body weight of rats in our previous study. Now we have determined the inhibitory characteristics and kinetic parameters of extracts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service