Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43975

Supelco

Ethylmalonic acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(COOH)2
CAS Number:
Molecular Weight:
132.11
Beilstein:
774334
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

mp

112-114 °C (lit.)

application(s)

clinical testing

format

neat

SMILES string

CCC(C(O)=O)C(O)=O

InChI

1S/C5H8O4/c1-2-3(4(6)7)5(8)9/h3H,2H2,1H3,(H,6,7)(H,8,9)

InChI key

UKFXDFUAPNAMPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylmalonic acid (EMA) is a branched fatty acid. It is a urinary organic acid, used as an important biochemical indicator of abnormal metabolism created by various diseases.

Application

Ethylmalonic acid may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Human biological fluids using liquid chromatography coupled to mass spectrometry (LC-MS).
  • Urine samples of uterine myoma and uterine cervical cancer patients using gas chromatography and gas chromatography–mass spectrometry.

It may be used as an analytical standard to investigate its role in the inhibition of succinate and malate transport through the mitochondrial membranes in rats.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas chromatographic profiling and pattern recognition analysis of urinary organic acids from uterine myoma patients and cervical cancer patients
Kim R-K, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 712(1-2), 11-22 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service