Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

40501

Supelco

Deacylgymnemic acid

analytical standard

Synonym(s):

(3β,4α,16β-21β,22α)-16,21,22,23,28-Pentahydroxyolean-12-en-3-yl β-D-glucopyranosiduronic acid, 3-O-β-D-Glucuronopyranosyl gymnemagenin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H58O12
CAS Number:
Molecular Weight:
682.84
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

CC1(C)C[C@H]2C3=CC[C@@H]4[C@@]5(C)CC[C@H](O[C@@H]6O[C@@H]([C@@H](O)[C@H](O)[C@H]6O)C(O)=O)[C@@](C)(CO)[C@@H]5CC[C@@]4(C)[C@]3(C)C[C@H](O)[C@@]2(CO)[C@@H](O)[C@@H]1O

InChI

1S/C36H58O12/c1-31(2)13-18-17-7-8-20-32(3)11-10-22(47-30-25(42)23(40)24(41)26(48-30)29(45)46)33(4,15-37)19(32)9-12-34(20,5)35(17,6)14-21(39)36(18,16-38)28(44)27(31)43/h7,18-28,30,37-44H,8-16H2,1-6H3,(H,45,46)/t18-,19+,20+,21-,22-,23-,24-,25+,26-,27-,28-,30+,32-,33-,34+,35+,36-/m0/s1

InChI key

NXUZSZLFZAMZLC-SOLHJDTNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Suzuki et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 113(4), 316-320 (1993-04-01)
A method of the quantitative analysis was established for the determination of deacylgymnemic acid (DAGA) in the alkaline hydrolysate of the sample containing gymnemic acids which are ingredients of Gymnema sylvestre R. BR. leaves, by means of high-performance liquid chromatography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service