Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

36188

Supelco

Pebulate

PESTANAL®, analytical standard

Synonym(s):

Propyl ethyl-n-butylthiolcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H21NOS
CAS Number:
Molecular Weight:
203.34
Beilstein:
1933846
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCCN(CC)C(=O)SCCC

InChI

1S/C10H21NOS/c1-4-7-8-11(6-3)10(12)13-9-5-2/h4-9H2,1-3H3

InChI key

SGEJQUSYQTVSIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The inhibition of fatty acid elongation by a thiocarbamate herbicide and its sulphoxide.
P B Barrett et al.
Biochemical Society transactions, 21(2), 184S-184S (1993-05-01)
Plant fatty acid elongation: sensitivity to thiocarbamate herbicides and their sulphoxides.
P B Barrett et al.
Biochemical Society transactions, 22(3), 260S-260S (1994-08-01)
A Baldwin et al.
Biochemical Society transactions, 28(6), 650-651 (2001-02-15)
The thiocarbamate herbicide pebulate inhibits fatty acid elongation, which is necessary for surface lipid biosynthesis. As both barley and wild oats are susceptible to pebulate, the safener dichlormid was used to study the reversal of its herbicidal effect. Fatty acid
Adam Baldwin et al.
Journal of experimental botany, 54(385), 1289-1294 (2003-03-26)
The thiocarbamates, such as pebulate (S-propyl butyl (ethyl) thiocarbamate) are a well-established class of herbicides. They inhibit fatty acid elongation, which is necessary for the biosynthesis of constituents of surface waxes and suberin and this has been proposed to be
C L Stiles et al.
Journal of agricultural and food chemistry, 48(10), 4681-4686 (2000-10-29)
Metam sodium is a potential replacement for methyl bromide, which is used to control soil pests. Metam sodium rapidly breaks down in the soil to form methylisothiocyanate (MITC). Dissipation of the herbicides EPTC and pebulate in a silt loam soil

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service